Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Hexadiyne 98.0+%, TCI America™
SDP

Catalog No. H05021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05021G 1 g
H05025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05021G Supplier TCI America Supplier No. H05021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2809-69-0
Molecular Formula C6H6
Molecular Weight (g/mol) 78.11
MDL Number MFCD00041617
InChI Key PCTCNWZFDASPLA-UHFFFAOYSA-N
Synonym 2,4-hexadiyne, dimethylbutadiyne, dimethyldiacetylene, acmc-209h0v, ch3c.$.cc.$.cch3, hexa-2,4-diyn-1-yl
PubChem CID 137727
IUPAC Name hexa-2,4-diyne
SMILES CC#CC#CC

Specifications

Melting Point 66°C
Boiling Point 129°C
Quantity 1 g
UN Number 1325
Formula Weight 78.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Hexadiyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.