Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,4-Lutidine, 98+%
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -60°C |
| Density | 0.931 |
| Boiling Point | 158°C to 160°C |
| Flash Point | 47°C (117°F) |
| Odor | Unpleasant |
| Refractive Index | 1.5 |
| Quantity | 25 g |
| UN Number | UN1992 |
| Formula Weight | 107.16 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H226-H301-H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H331-H226-H312-H315-H319-H335 Toxic if swallowed. Toxic if inhaled. Flammable liquid and vapour. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S. |
| EINECSNumber | 203-586-8 |
| RTECSNumber | OK9400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction