Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pentadienoic acid, 97%, stabilized

Catalog No. AC455740010
Click to view available options
Quantity:
1 g
5 g

CAS: 626-99-3 | C5H6O2 | 98.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-99-3
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
InChI Key SDVVLIIVFBKBMG-ONEGZZNKSA-N
Synonym penta-2,4-dienoic acid, 2,4-pentadienoic acid, trans-2,4-pentadienoic acid, 1,3-butadiene-1-carboxylic acid, e-2,4-pentadienoic acid, e-penta-2,4-dienoic acid, 2e-2,4-pentadienoic acid, beta-vinyl acrylic acid, vynilacrylic acid, 2e-penta-2,4-dienoic acid
PubChem CID 642034
ChEBI CHEBI:37331
IUPAC Name (2E)-penta-2,4-dienoic acid
SMILES C=CC=CC(=O)O

Specifications

Melting Point 69.0°C to 72.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula CH2(CH)3COOH
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2,4-Pentadienoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN:
EINECSNumber 210-976-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.