Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pentanedione, 99%

Catalog No. aaa14117ap
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 123-54-6 | C5H8O2 | 100.12 g/mol

2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

Solubility
It is readily miscible with water.

Notes
Incompatible with isocyanates, aldehydes, cyanides, peroxides, and anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O

Specifications

Melting Point -23°C
Density 0.973
Boiling Point 140°C to 141°C
Flash Point 34°C (93°F)
Assay Percent Range 99%
Refractive Index 1.452
Quantity 500 mL
UN Number 2310
Beilstein 741937
Merck Index 14,81
Solubility Information It is readily miscible with water.
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4-Pentanedione
Show More Show Less
Hazard Category H226-H302-H311+H331
Hazard Statement GHS H Statement
H311-H331-H226-H302
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANE-2,4-DIONE
EINECSNumber 204-634-0
RTECSNumber SA1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.