Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-pyridyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™

Catalog No. CC34701CB
Click to view available options
Quantity:
250 mg
1 g
10 g

Catalog No. CC34701CB

Supplier: Thermo Scientific Chemicals CC34701CB

Only null left
Add to Cart

Chemical Identifiers

CAS 21278-86-4
Molecular Formula C9H5N2O2S
Molecular Weight (g/mol) 205.21
MDL Number MFCD00171745
InChI Key COOQMBOJAAZEIR-UHFFFAOYSA-M
Synonym 2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole
PubChem CID 716091
IUPAC Name 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid
SMILES [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 206.22
Percent Purity 95%
Chemical Name or Material 2-(4-pyridyl)-1, 3-thiazole-4-carboxylic Acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.