Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole 98.0+%, TCI America™
SDP

Catalog No. B176725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B176725G 25 g
B17675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B176725G Supplier TCI America Supplier No. B176725G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 15082-28-7
Molecular Formula C24H22N2O
Molecular Weight (g/mol) 354.453
MDL Number MFCD00003101
InChI Key XZCJVWCMJYNSQO-UHFFFAOYSA-N
Synonym butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole
PubChem CID 84782
IUPAC Name 2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole
SMILES CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 25 g
Formula Weight 354.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.