Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine 98.0+%, TCI America™
SDP

Catalog No. A23081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23081G 1 g
A23085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23081G Supplier TCI America Supplier No. A23081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 152460-10-1
Molecular Formula C16H15N5
Molecular Weight (g/mol) 277.331
MDL Number MFCD09028125
InChI Key QGAIPGVQJVGBIA-UHFFFAOYSA-N
Synonym N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine
PubChem CID 9965429
IUPAC Name 4-methyl-3-N-(4-pyridin-3-ylpyrimidin-2-yl)benzene-1,3-diamine
SMILES CC1=C(C=C(C=C1)N)NC2=NC=CC(=N2)C3=CN=CC=C3

Specifications

Melting Point 145°C
Color Yellow
Quantity 1 g
Formula Weight 277.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.