Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone 95.0+%, TCI America™
SDP

Catalog No. B1839500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1839500G 500 g
B183925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1839500G Supplier TCI America Supplier No. B1839500G

Chemical Identifiers

CAS 903-19-5
Molecular Formula C22H38O2
Molecular Weight (g/mol) 334.544
MDL Number MFCD00128797
InChI Key CLDZVCMRASJQFO-UHFFFAOYSA-N
PubChem CID 70180
IUPAC Name 2,5-bis(2,4,4-trimethylpentan-2-yl)benzene-1,4-diol
SMILES CC(C)(C)CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)CC(C)(C)C)O

Specifications

Melting Point 130°C
Quantity 500 g
Formula Weight 334.54
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone
EINECSNumber (3)-0553
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.