Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile 95.0+%, TCI America™
SDP

Catalog No. B54581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54581G 1 g
B5458200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54581G Supplier TCI America Supplier No. B54581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 704890-84-6
Molecular Formula C12H5BrN2S2
Molecular Weight (g/mol) 321.21
MDL Number MFCD06214120
InChI Key GZLRCTASJSGPHL-UHFFFAOYSA-N
Synonym 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]propanedinitrile
PubChem CID 85975548
IUPAC Name 2-({5'-bromo-[2,2'-bithiophen]-5-yl}methylidene)propanedinitrile
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C=C(C#N)C#N

Specifications

Melting Point 202°C
Quantity 1 g
Formula Weight 321.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]malononitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.