Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diamino-4,6-dichloropyrimidine 98.0+%, TCI America™
SDP

Catalog No. D42311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42311G 1 g
D42315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42311G Supplier TCI America Supplier No. D42311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55583-59-0
Molecular Formula C4H4Cl2N4
Molecular Weight (g/mol) 179.004
MDL Number MFCD00792652
InChI Key ZXWGHENZKVQKPX-UHFFFAOYSA-N
PubChem CID 301039
IUPAC Name 4,6-dichloropyrimidine-2,5-diamine
SMILES C1(=C(N=C(N=C1Cl)N)Cl)N

Specifications

Melting Point 181°C
Color Red-Yellow
Quantity 1 g
Formula Weight 179.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diamino-4,6-dichloropyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.