Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diaminopyridine, 98+%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
4318-76-7
C5H7N3
109.13
MIROPXUFDXCYLG-UHFFFAOYSA-N
2,5-diaminopyridine,2,5-pyridinediamine,pyridine, 2,5-diamino,diaminopyridine van,unii-h6olz060hi,h6olz060hi,da1,pyridine,5-diamino,2,5-diamino pyridine,2,5-diamino-pyridine
20314
pyridine-2,5-diamine
C1=CC(=NC=C1N)N
4318-76-7
C5H7N3
109.13
MIROPXUFDXCYLG-UHFFFAOYSA-N
2,5-diaminopyridine,2,5-pyridinediamine,pyridine, 2,5-diamino,diaminopyridine van,unii-h6olz060hi,h6olz060hi,da1,pyridine,5-diamino,2,5-diamino pyridine,2,5-diamino-pyridine
20314
pyridine-2,5-diamine
C1=CC(=NC=C1N)N
Packaging Glass bottle
Boiling Point 180.0°C (12.0 mmHg)
Quantity 1g
Assay Percent Range 98% min. (HPLC)
Formula Weight 109.13
Percent Purity 98+%
Chemical Name or Material 2, 5-Diaminopyridine

GHS Signal Word: Danger