Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diaminotoluene 98.0+%, TCI America™
SDP

Catalog No. D462825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D462825G 25 g
D46285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D462825G Supplier TCI America Supplier No. D462825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-70-5
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00035779
InChI Key OBCSAIDCZQSFQH-UHFFFAOYSA-N
Synonym 2-Methyl-1,4-phenylenediamine
PubChem CID 7252
ChEBI CHEBI:53619
IUPAC Name 2-methylbenzene-1,4-diamine
SMILES CC1=C(C=CC(=C1)N)N

Specifications

Melting Point 62°C
Boiling Point 168°C
Quantity 25 g
UN Number 2811
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diaminotoluene
EINECSNumber (3)-0126
RTECSNumber XS9700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.