Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromohydroquinone, 97%

Catalog No. p-7036959
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 14753-51-6 | C6H4Br2O2 | 267.904 g/mol

It is an important pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate.

Solubility
Soluble in methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14753-51-6
Molecular Formula C6H4Br2O2
Molecular Weight (g/mol) 267.904
MDL Number MFCD00192664
InChI Key VALXCIRMSIFPFN-UHFFFAOYSA-N
PubChem CID 280945
IUPAC Name 2,5-dibromobenzene-1,4-diol
SMILES C1=C(C(=CC(=C1Br)O)Br)O

Specifications

Melting Point 191°C to 194°C
Quantity 1 g
Synonym 2,5-dibromohydroquinone, 2,5-dibromo-1,4-benzenediol, 2,5-dibromohydroquine, 1,4-benzenediol,2,5-dibromo, 2,5-dibromo-benzene-1,4-diol, 1,4-benzenediol, 2,5-dibromo, 2,5-dibromo-1,4-dihydroxybenzene
Solubility Information Soluble in methanol
Formula Weight 267.9
Percent Purity 97%
Chemical Name or Material 2,5-Dibromohydroquinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.