Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromothiophene, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
3141-27-3
C4H2Br2S
241.93
MFCD00005420
KBVDUUXRXJTAJC-UHFFFAOYSA-N
thiophene, 2,5-dibromo,2,5-dibromo-thiophene,unii-f0033k8cxm,pubchem5512,thiophene,5-dibromo,acmc-1ckmh,2,5-bis bromanyl thiophene,ksc224g0j,2,5-dibromothiophene,bidd:gt0086
18453
2,5-dibromothiophene
C1=C(SC(=C1)Br)Br
3141-27-3
C4H2Br2S
241.93
MFCD00005420
KBVDUUXRXJTAJC-UHFFFAOYSA-N
thiophene, 2,5-dibromo,2,5-dibromo-thiophene,unii-f0033k8cxm,pubchem5512,thiophene,5-dibromo,acmc-1ckmh,2,5-bis bromanyl thiophene,ksc224g0j,2,5-dibromothiophene,bidd:gt0086
18453
2,5-dibromothiophene
C1=C(SC(=C1)Br)Br
Refractive Index 1.6270 to 1.63
Density 2.1400g/mL
Melting Point -6°C
Boiling Point 211°C
Flash Point 99°C
Specific Gravity 2.14
Packaging Glass bottle
Color Yellow to Brown
Infrared Spectrum Authentic
Quantity 100g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Beilstein 17, 33
Solubility Information Solubility in water: practically insoluble
Formula Weight 241.93
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 2, 5-Dibromothiophene
Show More Show Less

GHS H Statement:
Fatal if swallowed.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : Product darkens on storage or exposure to light