Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromothiophene, 95%

Catalog No. AC148685000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 3141-27-3 | C4H2Br2S | 241.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3141-27-3
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005420
InChI Key KBVDUUXRXJTAJC-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
PubChem CID 18453
IUPAC Name 2,5-dibromothiophene
SMILES BrC1=CC=C(Br)S1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -6°C
Color Yellow to Brown
Density 2.1400g/mL
Boiling Point 211°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.6270 to 1.63
Quantity 500 g
Beilstein 17, 33
Specific Gravity 2.14
Solubility Information Solubility in water: practically insoluble
Formula Weight 241.93
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2, 5-Dibromothiophene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF
EINECSNumber 221-547-3
Recommended Storage Product darkens on storage or exposure to light

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.