Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dichloroacetophenone, 96%, Thermo Scientific™

Catalog No. AC179420100
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 2476-37-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000607
InChI Key CYNFEPKQDJHIMV-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
PubChem CID 75587
IUPAC Name 1-(2,5-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Cl)Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 11.0°C to 13.0°C
Color Brown to Yellow
Density 1.3120g/mL
Boiling Point 118.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5604 to 1.5624
Linear Formula Cl2C6H3COCH3
Quantity 10 g
Specific Gravity 1.312
Formula Weight 189.04
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2', 5'-Dichloroacetophenone, 96%
Show More Show Less
EINECSNumber 219-605-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.