Learn More
2,5-Dichloroaniline, 99%
Description
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.
Solubility
Soluble in dilute hydrochloric acid.
Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 48°C to 52°C |
Density | 1.54 |
Boiling Point | 250°C to 251°C |
Flash Point | 139°C (282°F) |
Quantity | 2500 g |
UN Number | UN3442 |
Beilstein | 1447438 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in dilute hydrochloric acid. |
Formula Weight | 162.02 |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H373-H500 |
Hazard Statement | GHS H Statement H301-H311-H331-H373 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID |
EINECSNumber | 202-455-2 |
RTECSNumber | BX2610000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.