Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloroaniline 98.0+%, TCI America™
SDP

Catalog No. D0322100G
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 95-82-9
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007667
InChI Key AVYGCQXNNJPXSS-UHFFFAOYSA-N
Synonym 1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
PubChem CID 7262
ChEBI CHEBI:34245
IUPAC Name 2,5-dichloroaniline
SMILES NC1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 52°C
Boiling Point 251°C
Quantity 100 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloroaniline
EINECSNumber (3)-0261
RTECSNumber BX2610000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.