Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorobenzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D2244100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2244100G 100 g
D224425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2244100G Supplier TCI America Supplier No. D2244100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5402-73-3
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00007429
InChI Key BXCOSWRSIISQSL-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzene-1-sulfonyl chloride, 2,5-dichlorobenzenesulphonyl chloride, 2,5-dichlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2,5-dichloro, 2,5-dichloro-benzenesulfonyl chloride, 2,5-dichlorophenylsulfonyl chloride, 2,5-dichlorophenyl chlorosulfone, pubchem5116, acmc-209lcc, bxcoswrsiisqsl-uhfffaoysa
PubChem CID 79401
IUPAC Name 2,5-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Cl)S(=O)(=O)Cl)Cl

Specifications

Melting Point 37°C
Color White-Yellow
Quantity 100 g
UN Number 3261
Formula Weight 245.50
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 2,5-Dichlorobenzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.