Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorobenzoic acid, 97%

Catalog No. p-104290
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle

CAS: 50-79-3 | C7H4Cl2O2 | 191.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-79-3
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002416
InChI Key QVTQYSFCFOGITD-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dichloro, unii-kp7cmr0gsd, 2,5-dichlorobenzoicacid, kp7cmr0gsd, 2,5-dichloro-benzoic acid, ccris 8609, pubchem3685, 2,5dichlorobenzoic acid, wln: qvr bg eg, dsstox_cid_4978
PubChem CID 5784
IUPAC Name 2,5-dichlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=CC=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 157.0°C
Color White to Beige
Boiling Point 301.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Linear Formula Cl2C6H3CO2H
Quantity 10 g
Beilstein 09, 342
Solubility Information Solubility in water: 0.8g/L (14°C). Other solubilities: soluble in most polar solvents,insoluble in non-polar solvents
Formula Weight 191.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 5-Dichlorobenzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-065-7
RTECSNumber DG6825000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.