Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,5-Dichlorobenzoic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Melting Point | 154°C |
Color | White-Yellow |
Quantity | 100 g |
Formula Weight | 191.01 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2,5-Dichlorobenzoic Acid |
Safety and Handling
EINECSNumber | (3)-1410&(3)-1424 |
RTECSNumber | DG6825000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction