Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloropyridine, 98%, Thermo Scientific™

Catalog No. AC186140100
Click to view available options
Quantity:
10 g

Catalog No. AC186140100

Supplier: Thermo Scientific Chemicals 186140100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16110-09-1
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006239
InChI Key GCTFDMFLLBCLPF-UHFFFAOYSA-N
Synonym pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
PubChem CID 27685
IUPAC Name 2,5-dichloropyridine
SMILES ClC1=CC=C(Cl)N=C1

Specifications

Melting Point 58.0°C to 62.0°C
Boiling Point 190.0°C to 191.0°C
Flash Point 112°C
Linear Formula (C5H3N)Cl2
Quantity 10 g
Solubility Information Solubility in water: insoluble.
Formula Weight 147.99
Chemical Name or Material 2,5-Dichloropyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.