Alfa Aesar™ 2,5-Difluoroaniline, 98%

Manufacturer: Alfa Aesar™ A1027218

Quantity 50g

 View more versions of this product

Catalog No. AAA1027218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 367-30-6
    Molecular Formula C6H5F2N
    Formula Weight 129.11
    MDL Number MFCD00007651
    UN Number UN2941
    Beilstein 2802549
    Chemical Name or Material 2,5-Difluoroaniline

    Specifications

    Formula Weight 129.11
    Health Hazard 2 GHS H Statement
    H311-H331-H373-H227-H302-H315-H319
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Molecular Formula C6H5F2N
    CAS 367-30-6
    MDL Number MFCD00007651
    UN Number UN2941
    Beilstein 2802549
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
    Boiling Point 171° to 174°C
    Melting Point 13° to 14°C
    Quantity 50g
    Flash Point 68°C (154°F)
    Odor Characteristic
    Chemical Name or Material 2,5-Difluoroaniline
    Assay 98%

    Description

    Applications
    2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

    Solubility
    Not miscible in water.

    Notes
    Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.

    GHS H Statement
    H311-H331-H373-H227-H302-H315-H319
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit