Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluoroanisole 98.0+%, TCI America™
SDP

Catalog No. D38055G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 75626-17-4
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00042561
InChI Key HUDMAQLYMUKZOZ-UHFFFAOYSA-N
PubChem CID 2724989
IUPAC Name 1,4-difluoro-2-methoxybenzene
SMILES COC1=C(C=CC(=C1)F)F

Specifications

Color Yellow
Boiling Point 73°C
Quantity 5 g
UN Number 1993
Synonym 2,5-difluoroanisole, 2,5-difluoromethoxybenzene, benzene, 1,4-difluoro-2-methoxy, 2,5-difluoro-1-methoxybenzene, 2,5-difluoroanisol, pubchem3456, acmc-209oz7, ksc641o8p, 1,4-difluoro-2-methoxy-benzene, benzene,1,4-difluoro-2-methoxy
Formula Weight 144.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluoroanisole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.