Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzoyl chloride, 97+%

Catalog No. p-7080889
Click to view available options
Quantity:
5 g
25 g

CAS: 35730-09-7 | C7H3ClF2O | 176.547 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35730-09-7
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00009929
InChI Key RLRUKKDFNWXXRT-UHFFFAOYSA-N
Synonym 2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride
PubChem CID 588082
IUPAC Name 2,5-difluorobenzoyl chloride
SMILES C1=CC(=C(C=C1F)C(=O)Cl)F

Specifications

Density 1.425
Boiling Point 81°C to 83°C (14 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.515
Quantity 5 g
UN Number UN3265
Beilstein 2046666
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzoyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 252-701-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.