Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorophenol, 96%, Thermo Scientific™

Catalog No. ac241950050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2713-31-7
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00042501
InChI Key INXKVYFOWNAVMU-UHFFFAOYSA-N
Synonym phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464
PubChem CID 94952
IUPAC Name 2,5-difluorophenol
SMILES OC1=CC(F)=CC=C1F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 35°C to 42°C
Color Beige or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Linear Formula F2(C6H3)OH
Quantity 5 g
Beilstein 10, IV, 780
Formula Weight 130.1
Percent Purity ≥95%
Physical Form Crystalline Mass, Crystals, Needles or Chunks
Chemical Name or Material 2, 5-Difluorophenol, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.