Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane 95.0+%, TCI America™
SDP

Catalog No. D378525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D378525G 25 g
D37855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D378525G Supplier TCI America Supplier No. D378525G

Chemical Identifiers

CAS 55704-78-4
Molecular Formula C6H12O2S2
Molecular Weight (g/mol) 180.28
MDL Number MFCD00010121
InChI Key NHKIYYMFGJBOTK-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2,5-dihydroxy-1,4-dithiane, dimeric mercapto propanone, 2-mercaptopropanone, 1,4-dithiane-2,5-diol, 2,5-dimethyl, 2,5-dimethyl-2,5-dihydroxy-p-dithiane, 2,5-dihydroxy-2,5-dimethyl-1,4-dithiane, 2,5-dimethyl-p-dithiane-2,5-diol, 2,5-dimethyl-1,4 dithiane-2,5-diol, fema no. 3450, dimericmercaptopropanone
PubChem CID 62105
IUPAC Name 2,5-dimethyl-1,4-dithiane-2,5-diol
SMILES CC1(CSC(CS1)(C)O)O

Specifications

Melting Point 88°C
Color White
Quantity 25 g
Formula Weight 180.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.