Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dihydroxybenzaldehyde, 98+%

Click to view available options
Quantity:
1g
5g
25g
1194-98-5
C7H6O3
138.122
MFCD00003333
CLFRCXCBWIQVRN-UHFFFAOYSA-N
gentisaldehyde,benzaldehyde, 2,5-dihydroxy,gentisate aldehyde,unii-0q83hds90w,polymer,2-formylhydroquinone,pubchem2285,acmc-1btnd,2,5 dihydroxybenzaldehyde,benzaldehyde,5-dihydroxy
70949
CHEBI:28508
2,5-dihydroxybenzaldehyde
C1=CC(=C(C=C1O)C=O)O
1194-98-5
C7H6O3
138.122
MFCD00003333
CLFRCXCBWIQVRN-UHFFFAOYSA-N
gentisaldehyde,benzaldehyde, 2,5-dihydroxy,gentisate aldehyde,unii-0q83hds90w,polymer,2-formylhydroquinone,pubchem2285,acmc-1btnd,2,5 dihydroxybenzaldehyde,benzaldehyde,5-dihydroxy
70949
CHEBI:28508
2,5-dihydroxybenzaldehyde
C1=CC(=C(C=C1O)C=O)O
Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air.
Sensitivity Air Sensitive
Melting Point 97°C to 101°C
Quantity 5g
Beilstein 1363961
Solubility Information Soluble in water.
Formula Weight 138.12
Percent Purity ≥98%
Chemical Name or Material 2,5-Dihydroxybenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-789-6

TSCA : Yes

Recommended Storage : Ambient temperatures