Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydroxybenzaldehyde, 99%

Catalog No. p-4258957
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 1194-98-5 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-98-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003333
InChI Key CLFRCXCBWIQVRN-UHFFFAOYSA-N
Synonym gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy
PubChem CID 70949
ChEBI CHEBI:28508
IUPAC Name 2,5-dihydroxybenzaldehyde
SMILES OC1=CC=C(O)C(C=O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color Green-Khaki to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 5 g
Beilstein 08,244
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol and ether
Formula Weight 138.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dihydroxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 214-789-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.