Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydroxyphenyl(diphenyl)phosphine Oxide 97.0+%, TCI America™
SDP

Catalog No. D375525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D375525G 25 g
D37555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D375525G Supplier TCI America Supplier No. D375525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13291-46-8
Molecular Formula C18H15O3P
Molecular Weight (g/mol) 310.289
InChI Key LLOXZCFOAUCDAE-UHFFFAOYSA-N
PubChem CID 2783495
IUPAC Name 2-diphenylphosphorylbenzene-1,4-diol
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=C(C=CC(=C3)O)O

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 310.29
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxyphenyl(diphenyl)phosphine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.