Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydroxyterephthalic acid, 97%

Catalog No. AAH6488806
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH6488806

Supplier: Thermo Scientific Chemicals H6488806

Only null left
Add to Cart

CAS: 610-92-4 | C8H6O6 | 198.13 g/mol

Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in hot dimethylformamide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-92-4
Molecular Formula C8H6O6
Molecular Weight (g/mol) 198.13
MDL Number MFCD00132933
InChI Key OYFRNYNHAZOYNF-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid
PubChem CID 69131
IUPAC Name 2,5-dihydroxyterephthalic acid
SMILES C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O

Specifications

Melting Point >300°C
Color Gray to Green
Quantity 5 g
Beilstein 2696052
Solubility Information Soluble in hot dimethylformamide.
Formula Weight 198.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dihydroxyterephthalic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-239-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.