Learn More
2,5-Diiodothiophene, 99%
Description
2,5-Diiodothiophene is used in the preparation of oligothiophene films and maskless fabrication of periodic patterns of a conjugated polymer. It finds application in the fabrication of oligothiophene and polythiophene micropatterns.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-Diiodothiophene is used in the preparation of oligothiophene films and maskless fabrication of periodic patterns of a conjugated polymer. It finds application in the fabrication of oligothiophene and polythiophene micropatterns.
Solubility
Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 39°C to 43°C |
Boiling Point | 139°C to 140°C (15 mmHg) |
Quantity | 5 g |
Beilstein | 109915 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 335.93 |
Percent Purity | 99% |
Chemical Name or Material | 2,5-Diiodothiophene |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 210-915-9 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.