Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimercapto-1,3,4-thiadiazole, 97% (dry wt.), water <3%

Catalog No. aaa1261518
Click to view available options
Quantity:
50 g
250 g

CAS: 1072-71-5 | C2H2N2S3 | 150.23 g/mol

2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-71-5
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
MDL Number MFCD00003103
InChI Key BIGYLAKFCGVRAN-UHFFFAOYSA-N
Synonym 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i
PubChem CID 2723630
IUPAC Name 1,3,4-thiadiazolidine-2,5-dithione
SMILES S=C1NNC(=S)S1

Specifications

Melting Point ∼162°C (decomposition)
Quantity 50 g
Beilstein 113444
Solubility Information Soluble in water.
Formula Weight 150.24
Percent Purity 98%
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-014-1
RTECSNumber XI3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.