Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxy-4-methylbenzaldehyde, 97%

Catalog No. AAH6414006
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 4925-88-6 | C10H12O3 | 180.20 g/mol

Used in the preparation of phenylamine derivative as potential psychotomimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of phenylamine derivative as potential psychotomimetics.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4925-88-6
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD02253192
InChI Key LRSRTWLEJBIAIT-UHFFFAOYSA-N
Synonym 4-methyl-2,5-dimethoxybenzaldehyde, unii-238b3l4auk, 2,5-dimethoxy-4-methyl-benzaldehyde, benzaldehyde, 2,5-dimethoxy-4-methyl, 2,5-dimethoxy-p-tolualdehyde, p-tolualdehyde, 2,5-dimethoxy, who-dd, 2,5-dimethoxy-4-methylbenzaldehyde;, 2,5-dimethoxy-4-methyl-benzaldehyde;, #
PubChem CID 602019
IUPAC Name 2,5-dimethoxy-4-methylbenzaldehyde
SMILES COC1=CC(C)=C(OC)C=C1C=O

Specifications

Melting Point 82°C to 86°C
Color White
Quantity 5 g
Beilstein 1866592
Solubility Information Soluble in dichloromethane,dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 180.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dimethoxy-4-methylbenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.