Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dimethoxyacetophenone 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1201-38-3
C10H12O3
180.2
MFCD00008728
FAXUIYJKGGUCBO-UHFFFAOYSA-N
2',5'-dimethoxyacetophenone,1-2,5-dimethoxyphenyl ethanone,2,5-dimethoxyacetophenone,1-2,5-dimethoxyphenyl ethan-1-one,ethanone, 1-2,5-dimethoxyphenyl,1-2,5-dimethoxyphenyl-ethanone,2-acetyl-1,4-dimethoxybenzene,acetophenone, 2',5'-dimethoxy,1-acetyl-2,5-dimethoxybenzene,pubchem13430
70991
1-(2,5-dimethoxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)OC)OC
1201-38-3
C10H12O3
180.2
MFCD00008728
FAXUIYJKGGUCBO-UHFFFAOYSA-N
2',5'-dimethoxyacetophenone,1-2,5-dimethoxyphenyl ethanone,2,5-dimethoxyacetophenone,1-2,5-dimethoxyphenyl ethan-1-one,ethanone, 1-2,5-dimethoxyphenyl,1-2,5-dimethoxyphenyl-ethanone,2-acetyl-1,4-dimethoxybenzene,acetophenone, 2',5'-dimethoxy,1-acetyl-2,5-dimethoxybenzene,pubchem13430
70991
1-(2,5-dimethoxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)OC)OC
Quantity 100g
Melting Point 18°C to 20°C
Flash Point >112°C
Boiling Point 155°C to 158°C (11.0mmHg)
Specific Gravity 1.13
Density 1.1300g/mL
Refractive Index 1.5421 to 1.5441
Color Green-Yellow to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)2C6H3COCH3
Beilstein 08, 271
Formula Weight 180.2
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2', 5'-Dimethoxyacetophenone, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 214-858-0

TSCA : TSCA