Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxybenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. D1111250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1111250G 250 g
D111125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1111250G Supplier TCI America Supplier No. D1111250G

Chemical Identifiers

CAS 93-02-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003314
InChI Key AFUKNJHPZAVHGQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde
PubChem CID 66726
IUPAC Name 2,5-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)OC)C=O

Specifications

Melting Point 50°C
Boiling Point 270°C
Quantity 250 g
Formula Weight 166.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxybenzaldehyde
EINECSNumber (3)-2665
RTECSNumber CU5740500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.