Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxyphenylmagnesium bromide, 0.5M in 2-MeTHF

Catalog No. aah54337ae
Click to view available options
Quantity:
100 mL

CAS: 62890-98-6 | C8H9BrMgO2 | 241.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62890-98-6
Molecular Formula C8H9BrMgO2
Molecular Weight (g/mol) 241.37
MDL Number MFCD01311475
InChI Key GRSCPLVZIMBKKY-UHFFFAOYSA-M
Synonym 2,5-dimethoxyphenylmagnesium bromide, 2,5-dimethoxyphenyl magnesium bromide, 2,5-dimethoxyphenylmagnesium bromide solution, grscplvzimbkky-uhfffaoysa-m, 2,5-dimethoxylphenylmagnesium bromide, magnesium,1,4-dimethoxybenzene-6-ide,bromide, 2,5-dimethoxyphenylmagnesium bromide, 0.5m in 2-methf, 2,5-dimethoxyphenylmagnesium bromide 0.5 m in tetrahydrofuran, 2,5-dimethoxyphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 11085924
IUPAC Name magnesium(2+) 2,5-dimethoxybenzen-1-ide bromide
SMILES COC1=CC=C(OC)C([Mg]Br)=C1

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Formula Weight 241.37
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 2,5-Dimethoxyphenylmagnesium bromide
Hazard Category H225-H260-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.