Learn More
2,5-Dimethoxytetrahydrofuran, cis + trans, 98%
Description
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.
Solubility
Miscible with methanol, diethyl ether and tetrahydrofuran. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.

Specifications
Specifications
Melting Point | -45°C |
Density | 1.02 |
Boiling Point | 145°C to 147°C |
Quantity | 100 g |
Refractive Index | 1.418 |
UN Number | UN3271 |
Beilstein | 104261 |
Solubility Information | Miscible with methanol,diethyl ether and tetrahydrofuran. Immiscible with water. |
Formula Weight | 132.16 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H226-H319-H331 |
Hazard Statement | GHS H Statement H330-H226-H302-H315-H319-H335 Fatal if inhaled. Flammable liquid and vapor. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S. |
EINECSNumber | 211-797-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.