Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylaniline, 99%

Catalog No. p-115392
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

CAS: 95-78-3 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-78-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007743
InChI Key VOWZNBNDMFLQGM-UHFFFAOYSA-N
Synonym 2,5-xylidine, p-xylidine, 2,5-dimethylphenylamine, 2-amino-1,4-xylene, 2,5-dimethylbenzenamine, benzenamine, 2,5-dimethyl, 5-methyl-o-toluidine, 6-methyl-m-toluidine, 1-amino-2,5-dimethylbenzene, p-dimethylaniline
PubChem CID 7259
ChEBI CHEBI:518305
IUPAC Name 2,5-dimethylaniline
SMILES CC1=CC=C(C)C(N)=C1

Specifications

Melting Point 11.5°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9700g/mL
Boiling Point 218°C
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5582 to 1.5602
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.97
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1135
Merck Index 15, 10282
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,carbon tetrachloride
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethylaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-451-0
RTECSNumber ZE9100000
TSCA TSCA
Recommended Storage product darkens if exposed to light and air

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.