Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylbenzenesulfonyl chloride, 98%, Thermo Scientific™

Catalog No. p-7059543
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19040-62-1
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00024875
InChI Key FZVZUIBYLZZOEW-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f
PubChem CID 87910
IUPAC Name 2,5-dimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C=C1)C)S(=O)(=O)Cl

Specifications

Melting Point 20°C
Color Yellow
Boiling Point 120°C to 125°C
Quantity 1 g
Physical Form Liquid/Solid
UN Number UN3265
Formula Weight 204.68
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber DB8927600
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.