Learn More
2,5-Dimethylfuran, 98+%
Description
2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.
Solubility
Slightly miscible with water. Miscible with ethanol and fats.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Melting Point | -62°C |
| Density | 0.903 |
| Boiling Point | 92°C to 93°C |
| Flash Point | 1°C (33°F) |
| Refractive Index | 1.441 |
| Quantity | 100 g |
| UN Number | UN3271 |
| Beilstein | 106449 |
| Solubility Information | Slightly miscible with water. Miscible with ethanol and fats. |
| Formula Weight | 96.13 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H225-H301-H315-H319-H317-H335 Highly flammable liquid and vapour. Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S. |
| EINECSNumber | 210-914-3 |
| RTECSNumber | LU0875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.