Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylfuran 98.0+%, TCI America™
SDP

Catalog No. D0725100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0725100ML 100 mL
D072525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0725100ML Supplier TCI America Supplier No. D0725100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 625-86-5
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00003250
InChI Key GSNUFIFRDBKVIE-UHFFFAOYSA-N
Synonym furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg
PubChem CID 12266
IUPAC Name 2,5-dimethylfuran
SMILES CC1=CC=C(O1)C

Specifications

Color Yellow
Boiling Point 93°C
Quantity 100 mL
UN Number 1993
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylfuran
RTECSNumber LU0875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.