Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenyl isocyanate, 98%, Thermo Scientific™

Catalog No. AC366800010
Click to view available options
:
1g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40397-98-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H9NO
Linear Formula (CH3)C6H3NCO
MDL Number MFCD00013854
Synonym 2,5-dimethylphenyl isocyanate, 2,5-dimethylphenylisocyanate, benzene, 2-isocyanato-1,4-dimethyl, 2,5-dimethyl phenylisocyanate, 2,5-dimethylbenzenisocyanate, acmc-1art9, 1-isocyanato-2,5-dimethyl-benzene
InChI Key SOXVXJQIQVOCAY-UHFFFAOYSA-N
SMILES CC1=CC=C(C)C(=C1)N=C=O
IUPAC Name 2-isocyanato-1,4-dimethylbenzene
Molecular Weight (g/mol) 147.18
PubChem CID 98605
ChEBI CHEBI:60097
Formula Weight 147.18
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Dimethylphenyl isocyanate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is co
Recommended Storage Refrigerator +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.