Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. D427125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D427125G 25 g
D42715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D427125G Supplier TCI America Supplier No. D427125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16213-85-7
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00013816
InChI Key HCELORQTHSKCQL-UHFFFAOYSA-N
Synonym 2,5-dimethylphenylacetonitrile, 2-2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzyl cyanide, acetonitrile, 2,5-xylyl, 2,5-xylyl acetonitrile, benzeneacetonitrile, 2,5-dimethyl, 2,5-xylylacetonitrile, 2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzeneacetonitrile, unii-ult00p22mn
PubChem CID 27753
IUPAC Name 2-(2,5-dimethylphenyl)acetonitrile
SMILES CC1=CC(=C(C=C1)C)CC#N

Specifications

Melting Point 30°C
Color White
Boiling Point 79°C
Quantity 25 g
UN Number 3439
Formula Weight 145.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dimethylphenylacetonitrile
RTECSNumber AM2675000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.