Alfa Aesar™ 2,5-Dimethylpyrazine, 99%

Manufacturer: Alfa Aesar™ A1235206

Quantity 5g

 View more versions of this product

Catalog No. AAA1235206

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 123-32-0
    Molecular Formula C6H8N2
    Formula Weight 108.14
    MDL Number MFCD00006147
    Beilstein 107052
    Synonym Ketine
    Chemical Name or Material 2,5-Dimethylpyrazine

    Specifications

    Formula Weight 108.14
    CAS 123-32-0
    Molecular Formula C6H8N2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Odor Pleasant
    Quantity 5g
    MDL Number MFCD00006147
    Beilstein 107052
    Synonym Ketine
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    RTECS Number UQ2800000
    Boiling Point 154° to 155°C
    Melting Point 15°C
    TSCA TSCA
    Flash Point 63°C (145°F)
    Chemical Name or Material 2,5-Dimethylpyrazine
    Assay 99%

    GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit