Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylthiophene, 98.5%

Catalog No. AC116880250
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle

CAS: 638-02-8 | C6H8S | 112.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-02-8
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00005452
InChI Key GWQOOADXMVQEFT-UHFFFAOYSA-N
PubChem CID 12514
IUPAC Name 2,5-dimethylthiophene
SMILES CC1=CC=C(S1)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9840g/mL
Boiling Point 134°C to 136°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.514
Quantity 25 mL
Beilstein 17, 41
Specific Gravity 0.984
Synonym thiophene, 2,5-dimethyl, 2,5-dimethyl-thiophene, unii-v6ddx6wb12, v6ddx6wb12, pubchem5194, 2,5 dimethylthiophene, thiophene,5-dimethyl, 2,5-dimethyl thiophene, thiophene,2,5-dimethyl, 2,5-dimethyl-2h-thiolium
Solubility Information Solubility in water: insoluble. Other solubilities: in most org solvents
Formula Weight 112.19
Percent Purity 98.50%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethylthiophene, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 211-313-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.