Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Pyrazinedicarboxylic acid hydrate, 97%

Catalog No. ac292320010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

CAS: 205692-63-3 | C6H4N2O4·2H2O | 204.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205692-63-3
Molecular Formula C6H4N2O4·2H2O
Molecular Weight (g/mol) 204.14
MDL Number MFCD00216919
InChI Key KOUKXHPPRFNWPP-UHFFFAOYSA-N
Synonym pyrazine-2,5-dicarboxylic acid hydrate, 2,5-pyrazinedicarboxylic acid dihydrate
PubChem CID 73553845
IUPAC Name pyrazine-2,5-dicarboxylic acid;hydrate
SMILES C1=C(N=CC(=N1)C(=O)O)C(=O)O.O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 257°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,acetone,ethylacetate, slightly soluble in ethanol,ether,chloroform,, benzene,petr. ether
Formula Weight 204.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2,5-Pyrazinedicarboxylic acid dihydrate, 97%
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.