Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6,10,14-Tetramethylpentadecane, 95%, ACROS Organics™

Click to view available options
Quantity:
5mL
50mL
100mL
250mL
Packaging:
Glass bottle
1921-70-6
C19H40
268.51
XOJVVFBFDXDTEG-UHFFFAOYSA-N
pristane,norphytane,bute hydrocarbon,norphytan,pristan,pentadecane, 2,6,10,14-tetramethyl,meso-pristane,2,6,10,10-tetramethylpentadecane,meso-form,norphytane, robuoy
15979
CHEBI:53181
2,6,10,14-tetramethylpentadecane
CC(C)CCCC(C)CCCC(C)CCCC(C)C
1921-70-6
C19H40
268.51
XOJVVFBFDXDTEG-UHFFFAOYSA-N
pristane,norphytane,bute hydrocarbon,norphytan,pristan,pentadecane, 2,6,10,14-tetramethyl,meso-pristane,2,6,10,10-tetramethylpentadecane,meso-form,norphytane, robuoy
15979
CHEBI:53181
2,6,10,14-tetramethylpentadecane
CC(C)CCCC(C)CCCC(C)CCCC(C)C
Boiling Point 296°C
Refractive Index 1.4369 to 1.4389
Flash Point >110°C
Specific Gravity 0.78
Viscosity 5 mPa.s (25°C)
Packaging Glass bottle
Density 0.7800g/mL
Infrared Spectrum Authentic
Color Colorless
Quantity 250mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)(CH2)3CH(CH3)2
Merck Index 15,787
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,petroleum ether,benzene,,chloroform and carbon tetrachloride
Formula Weight 268.51
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 2,6,10,14-Tetramethylpentadecane, 96%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 217-650-8

RTECSNumber : RZ1880000

TSCA : TSCA