Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis[(2S,4S)-4-methyl-5,5-diphenyloxazolidin-2-yl]pyridine, TCI America™
SDP

Catalog No. B422750MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
50 mg

Chemical Identifiers

CAS 1450841-25-4
Molecular Formula C37H35N3O2
Molecular Weight (g/mol) 553.706
InChI Key RQXPVLMMPSKAED-AIYQPOGASA-N
PubChem CID 72192467
IUPAC Name (2S,4S)-4-methyl-2-[6-[(2S,4S)-4-methyl-5,5-diphenyl-1,3-oxazolidin-2-yl]pyridin-2-yl]-5,5-diphenyl-1,3-oxazolidine
SMILES CC1C(OC(N1)C2=NC(=CC=C2)C3NC(C(O3)(C4=CC=CC=C4)C5=CC=CC=C5)C)(C6=CC=CC=C6)C7=CC=CC=C7

Specifications

Color White-Yellow
Quantity 50 mg
Formula Weight 553.71
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis[(2S,4S)-4-methyl-5,5-diphenyloxazolidin-2-yl]pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.