Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(4-fluorobenzylidene)cyclohexanone 96.0+%, TCI America™
SDP

Catalog No. B228225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B228225G 25 g
B22825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B228225G Supplier TCI America Supplier No. B228225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62085-74-9
Molecular Formula C20H16F2O
Molecular Weight (g/mol) 310.344
MDL Number MFCD00112608
InChI Key STPNBQOTKFPYEG-UNZYHPAISA-N
Synonym 2,6-Bis(4-fluorobenzal)cyclohexanone
PubChem CID 1551384
IUPAC Name (2E,6E)-2,6-bis[(4-fluorophenyl)methylidene]cyclohexan-1-one
SMILES C1CC(=CC2=CC=C(C=C2)F)C(=O)C(=CC3=CC=C(C=C3)F)C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 310.34
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(4-fluorobenzylidene)cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.